Novus Biologicals
Manufacturer Code:NBP303786100UL
Catalog # NB094862
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 660-800 of human Liprin alpha 1 (NP_003617.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ42630 FLJ43474 LAR-interacting protein 1 LIP.1 LIP-1 LIP1FLJ41337 LIPRIN liprin-alpha-1 Liprin-alpha1 MGC26800 Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-1 protein tyrosine phosphatase receptor type f polypeptide (PTPRF) interactingprotein (liprin) alpha 1 PTPRF-interacting protein alpha-1; LAR-interacting protein 1; LIP-1; Liprin-alpha-1; Liprin-alpha1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; PTPRF-interacting protein alpha-1
Gene Aliases: LIP.1; LIP1; LIPRIN; PPFIA1
UniProt ID: (Human) Q13136
Entrez Gene ID: (Human) 8500
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.