Proteintech
Manufacturer Code:141751AP150UL
Catalog # 50-172-9265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Liprin alpha 1 Fusion Protein Ag5381 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAR interacting protein 1 LIP 1 LIP.1 LIP1 LIPRIN Liprin alpha 1 PPFIA1; LAR-interacting protein 1; LIP-1; liprin; Liprin-alpha-1; Liprin-alpha1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein, alpha 1; PTPRF-interacting protein alpha-1
Gene Aliases: C030014K08Rik; C87158; LIP.1; LIP1; LIPRIN; PPFIA1
UniProt ID: (Human) Q13136
Entrez Gene ID: (Human) 8500, (Rat) 293645, (Mouse) 233977
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.