Novus Biologicals
Manufacturer Code:NBP271178APC
Catalog # NB271178APC
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A10 |
Immunogen | Recombinant protein fragment corresponding to amino acids 28-475 of human Lipoprotein Lipase/LPL (NP_000228) produced in HEK293T. |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.1.1 HDLCQ11 LIPDEC 3.1.1.34 lipoprotein lipase; Lipoprotein lipase; LPL; Phospholipase A1
Gene Aliases: HDLCQ11; LIPD; LPL
UniProt ID: (Human) P06858
Entrez Gene ID: (Human) 4023
Molecular Function:
esterase
hydrolase
lipase
storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.