Novus Biologicals
Manufacturer Code:NBP219364
Catalog # NBP219364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human LPL. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1 HDLCQ11 LIPDEC 3.1.1.34 lipoprotein lipase; Lipoprotein lipase; LPL; Phospholipase A1
Gene Aliases: HDLCQ11; LIPD; LPL
UniProt ID: (Human) P06858
Entrez Gene ID: (Human) 4023
Molecular Function: esterase hydrolase lipase storage protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.