Novus Biologicals
Manufacturer Code:NBP2852140.1ML
Catalog # NB033143
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of human Lipolysis Stimulated Lipoprotein Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ILDR3 immunoglobulin-like domain containing receptor 3 lipolysis stimulated lipoprotein receptor lipolysis-stimulated lipoprotein receptor lipolysis-stimulated receptor lipolysis-stimulated remnant LISCH LISCH protein LISCH7 liver-specific bHLH-Zip transcription factor MGC10659 MGC48312 MGC48503; immunoglobulin-like domain containing receptor 3; Lipolysis-stimulated lipoprotein receptor; lipolysis-stimulated remnant; LISCH protein; liver-specific bHLH-Zip transcription factor
Gene Aliases: ILDR3; LISCH; LISCH7; LSR
UniProt ID: (Human) Q86X29
Entrez Gene ID: (Human) 51599
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.