Novus Biologicals
Manufacturer Code:NBP271726AF647
Catalog # N271726A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI8F10 |
Immunogen | Full length recombinant protein of human Lipin 1 (NP_663731) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp781P1796 EC 3.1.3.18 EC 3.1.3.4 EC 5.4.2.7 KIAA0188phosphatidate phosphatase LPIN1 lipin 1 lipin-1 PAP1; Lipin-1; Phosphatidate phosphatase LPIN1
Gene Aliases: KIAA0188; LPIN1; PAP1
UniProt ID: (Human) Q14693
Entrez Gene ID: (Human) 23175
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.