Proteintech
Manufacturer Code:161781AP150UL
Catalog # 50-173-0772
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CSDD2 lin 28 homolog B (C. elegans) Lin 28B Lin28 LIN28B Protein lin 28 homolog B; Lin-28.2; Lin-28B; Protein lin-28 homolog B
Gene Aliases: 2810403D23Rik; CSDD2; D030047M17Rik; Lin-28.2; LIN28B
UniProt ID: (Human) Q6ZN17, (Mouse) Q45KJ6
Entrez Gene ID: (Human) 389421, (Mouse) 380669
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.