Proteintech
Manufacturer Code:161771AP150UL
Catalog # 50-173-0771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CSDD1 LIN 28 lin 28 homolog (C. elegans) Lin 28A LIN28 Lin28A Protein lin 28 homolog A ZCCHC1; Lin-28A; Protein lin-28 homolog A; RNA-binding protein LIN-28; testis expressed gene 17; Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1; zinc finger, CCHC domain containing 1
Gene Aliases: AL024421; CSDD1; ENSMUSG00000070700; Gm10299; LIN-28; lin-28A; LIN28; LIN28A; RGD1566408; Tex17; ZCCHC1
UniProt ID: (Human) Q9H9Z2, (Mouse) Q8K3Y3
Entrez Gene ID: (Human) 79727, (Rat) 500562, (Mouse) 83557
Molecular Function: DNA binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.