Invitrogen
Manufacturer Code:PA518522
Catalog # PIPA518522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TQDDIEKHQSDAG) corresponding to the internal amino acids of LEPR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B219; CD295; HuB219; LEP-R; Leptin receptor; Leptin receptor (fatty); leptin receptor gene-related protein; OB receptor; OB-R; OB-R LEP-R OB receptor HuB219 CD295 antigen LEPR leptin receptor CD295 OBR OB receptor LeprB
Gene Aliases: CD295; DB; diabetes; Fa; LEP-R; LEPR; Leprb; LEPRD; LEPROT; Modb1; OB-R; OB-RGRP; obese-like; obl; OBR
UniProt ID: (Human) P48357, (Rat) Q62959, (Mouse) P48356
Entrez Gene ID: (Human) 3953, (Rat) 24536, (Mouse) 16847
Molecular Function: cytokine defense/immunity protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.