Invitrogen
Manufacturer Code:PA128844
Catalog # PIPA128844
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment - amino acids 217-376 of Mouse Leptin Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B219; CD295; LEP-R; Leptin receptor; leptin receptor gene-related protein; OB receptor; OB-R; OB-R LEP-R OB receptor HuB219 CD295 antigen
Gene Aliases: Db; diabetes; Lepr; Leprb; LEPROT; Modb1; OB-RGRP; obese-like; obl; Obr
UniProt ID: (Mouse) P48356
Entrez Gene ID: (Mouse) 16847
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.