Novus Biologicals
Manufacturer Code:NBP121725IR
Catalog # NBP121725IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0085-9C29 |
Immunogen | Mouse recombinant EC-domain of LeptinR |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rat Monoclonal Antibody
Protein Aliases: CD295; CD295 antigen DB DKFZp686B1731 huB219 LEP-R leptin receptor OB receptor OB-R OBRCD295; HuB219; LEP-R; Leptin receptor; OB receptor; OB-R
Gene Aliases: CD295; DB; LEP-R; LEPR; LEPRD; OB-R; OBR
UniProt ID: (Human) P48357
Entrez Gene ID: (Human) 3953
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.