R&D Systems
Manufacturer Code:AF389SP
Catalog # AF389SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Leptin R Thr20-Asp839 Accession # P48357 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: B219 CD295 CD295 antigen DB DKFZp686B1731 huB219 LEPR LEP-R leptin receptor LeptinR OB R OB receptor OB-R OBRCD295; CD295; HuB219; LEP-R; Leptin receptor; OB receptor; OB-R
Gene Aliases: CD295; DB; LEP-R; LEPR; LEPRD; OB-R; OBR
UniProt ID: (Human) P48357
Entrez Gene ID: (Human) 3953
Molecular Function:
cytokine
defense/immunity protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.