Novus Biologicals
Manufacturer Code:NBP221619AF532
Catalog # NP221619A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6 |
Immunogen | Recombinant human leptin adsorbed onto aluminium hydroxide gel. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ94114 leptin leptin (murine obesity homolog) leptin (obesity homolog mouse) Obese protein obese mouse homolog of Obesity factor OBOBS; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); Obese protein; obese, mouse, homolog of; Obesity factor
Gene Aliases: LEP; LEPD; OB; OBS
UniProt ID: (Human) P41159
Entrez Gene ID: (Human) 3952
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.