Novus Biologicals
Manufacturer Code:NBP221996AF594
Catalog # NP221996A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 46I3 |
Immunogen | This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant Legumain. Mouse anti human Legumain Q99538. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AEP Asparaginyl endopeptidase cysteine protease 1 legumain LGMN1 Protease cysteine 1 protease cysteine 1 (legumain) PRSC1EC 3.4.22.34; Asparaginyl endopeptidase; cysteine protease 1; Legumain; Protease, cysteine 1; protease, cysteine, 1 (legumain)
Gene Aliases: AEP; LGMN; LGMN1; PRSC1
UniProt ID: (Human) Q99538
Entrez Gene ID: (Human) 5641
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.