R&D Systems
Manufacturer Code:AF2199
Catalog # AF2199
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Legumain/Asparaginyl Endopeptidase Ile18-Tyr433 Accession # Q99538 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AEP Asparaginyl endopeptidase cysteine protease 1 legumain LGMN LGMN1 Protease cysteine 1 protease cysteine 1 (legumain) PRSC1EC 3.4.22.34; Asparaginyl endopeptidase; cysteine protease 1; Legumain; Protease, cysteine 1; protease, cysteine, 1 (legumain)
Gene Aliases: AEP; LGMN; LGMN1; PRSC1
UniProt ID: (Human) Q99538
Entrez Gene ID: (Human) 5641
Molecular Function: cysteine protease hydrolase protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.