Abnova
Manufacturer Code:PAB10257
Catalog # 89-114-731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 361-373 of mouse Ldb1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Ldb1.
For Research Use Only
Protein Aliases: Carboxyl-terminal LIM domain-binding protein 2; CLIM-2; CLIM2; LDB-1; LIM domain-binding factor CLIM2; LIM domain-binding protein 1; mLdb1; NLI; Nuclear LIM interactor
Gene Aliases: CLIM2; Ldb1; NLI
UniProt ID: (Mouse) P70662
Entrez Gene ID: (Mouse) 16825
Molecular Function:
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.