Novus Biologicals
Manufacturer Code:NBP185804
Catalog # NBP185804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:MGCGCSSHPEDDWMENIDVCENCHYPIVPLDGKGTLLIRNGSEVRDPLVTYEGSNPPASPLQDNLVIALHSYEPSHDGDLGFEKGEQLRILEQSGEWWKAQSLTTGQEGFIPFNFVAKANSLEPEPWFFKNLSRKDAERQLLAPGNTHGS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.2 Leukocyte C-terminal Src kinase LSK Lymphocyte cell-specific protein-tyrosine kinase lymphocyte-specific protein tyrosine kinase p56(LSTRA) protein-tyrosine kinase p56lck p56-LCK pp58lck Protein YT16 Proto-oncogene Lck proto-oncogene tyrosine-protein kinase LCK T cell-specific protein-tyrosine kinase T-lymphocyte specific protein tyrosine kinase p56lck tyrosine-protein kinase Lck YT16; Leukocyte C-terminal Src kinase; LSK; Lymphocyte cell-specific protein-tyrosine kinase; p56(LSTRA) protein-tyrosine kinase; p56-LCK; Protein YT16; Proto-oncogene Lck; proto-oncogene tyrosine-protein kinase LCK; T cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; Tyrosine-protein kinase Lck
Gene Aliases: IMD22; LCK; LSK; p56lck; pp58lck; YT16
UniProt ID: (Human) P06239
Entrez Gene ID: (Human) 3932
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.