R&D Systems
Manufacturer Code:AF3704SP
Catalog # AF3704SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Lck Met1-Val66 Accession # P06239 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.2 Hck-3 Lcktkr Leukocyte C-terminal Src kinase LSK LSTRA Lymphocyte cell-specific protein-tyrosine kinase lymphocyte-specific protein tyrosine kinase p56(LSTRA) protein-tyrosine kinase p56lck p56-LCK pp58lck Protein YT16 Proto-oncogene Lck proto-oncogene tyrosine-protein kinase LCK T cell-specific protein-tyrosine kinase T-lymphocyte specific protein tyrosine kinase p56lck tyrosine-protein kinase Lck YT16; Leukocyte C-terminal Src kinase; LSK; Lymphocyte cell-specific protein-tyrosine kinase; p56(LSTRA) protein-tyrosine kinase; p56-LCK; Protein YT16; Proto-oncogene Lck; proto-oncogene tyrosine-protein kinase LCK; T cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; Tyrosine-protein kinase Lck
Gene Aliases: IMD22; LCK; LSK; p56lck; pp58lck; YT16
UniProt ID: (Human) P06239
Entrez Gene ID: (Human) 3932
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.