Abnova
Manufacturer Code:PAB12121
Catalog # 89-115-619
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant His fusion protein corresponding to C-terminus of mouse Lcat. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Lcat.
For Research Use Only
Protein Aliases: 1-alkyl-2-acetylglycerophosphocholine esterase; AI046659; D8Wsu61e; Lecithin-cholesterol acyltransferase; lecithin-cholesterol acyltransferase Lcat; PAF acetylhydrolase; Phosphatidylcholine-sterol acyltransferase; Phospholipid-cholesterol acyltransferase; Platelet-activating factor acetylhydrolase
Gene Aliases: AI046659; D8Wsu61e; Lcat
UniProt ID: (Mouse) P16301
Entrez Gene ID: (Mouse) 16816
Molecular Function: acyltransferase hydrolase lipase phospholipase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.