Novus Biologicals
Manufacturer Code:NBP223723SS
Catalog # NBP223723SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amino acids 70-86 CTAEVLYPSTGQETAPE of human Latexin protein were used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ECI; ECIMUM Endogenous carboxypeptidase inhibitor latexin TCIProtein MUM Tissue carboxypeptidase inhibitor; Endogenous carboxypeptidase inhibitor; Latexin; MUM; Protein MUM; TCI; Tissue carboxypeptidase inhibitor
Gene Aliases: ECI; LXN; TCI
UniProt ID: (Human) Q9BS40
Entrez Gene ID: (Human) 56925
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.