Novus Biologicals
Manufacturer Code:NBP243671
Catalog # NBP243671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human LTBP4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: latent transforming growth factor beta binding protein 4 LTBP-4 LTBP4L LTBP4S; latent transforming growth factor-beta binding protein 4L; Latent-transforming growth factor beta-binding protein 4; LTBP-4
Gene Aliases: ARCL1C; LTBP-4; LTBP4; LTBP4L; LTBP4S
UniProt ID: (Human) Q8N2S1
Entrez Gene ID: (Human) 8425
Molecular Function: annexin calcium-binding protein calmodulin cell adhesion molecule extracellular matrix glycoprotein extracellular matrix protein extracellular matrix structural protein intracellular calcium-sensing protein signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.