Novus Biologicals
Manufacturer Code:NBP211783FR
Catalog # NBP211783FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0447-5L38 |
Immunogen | Human LTBP1 |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: latent transforming growth factor beta binding protein 1 latent-transforming growth factor beta-binding protein 1 LTBP-1 MGC163161 TGF-beta1-BP-1 Transforming growth factor beta-1-binding protein 1; Latent-transforming growth factor beta-binding protein 1; LTBP-1; TGF-beta1-BP-1; Transforming growth factor beta-1-binding protein 1
Gene Aliases: LTBP1
UniProt ID: (Human) Q14766
Entrez Gene ID: (Human) 4052
Molecular Function:
annexin
calcium-binding protein
calmodulin
cell adhesion molecule
extracellular matrix glycoprotein
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.