Invitrogen
Manufacturer Code:PA5113036
Catalog # PIPA5113036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Ceramide synthase 6 protein (324-392AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4732462C07Rik AW544719 Ceramide synthase 6 CERS5 CERS6 LAG1 homolog ceramide synthase 6 LAG1 longevity assurance homolog 6 LASS6 longevity assurance homolog 6 MGC129949 MGC129950 RGD1561599 T1L; Ceramide synthase 6; CerS6; LAG1 homolog, ceramide synthase 6; LAG1 longevity assurance homolog 6; longevity assurance homolog 6
Gene Aliases: CERS5; CERS6; LASS6
UniProt ID: (Human) Q6ZMG9
Entrez Gene ID: (Human) 253782
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.