Novus Biologicals
Manufacturer Code:NBP177328
Catalog # NBP177328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human LASS5. The immunogen is located within the last 50 amino acids of LASS5. Amino Acid Squence: SKDDRSDVESSSEEEDV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 5; CerS5; CerS5 FLJ25304 LAG1 homolog ceramide synthase 5 LAG1 longevity assurance homolog 5 LAG1 longevity assurance homolog 5 (S. cerevisiae) MGC45411 Trh4; LAG1 homolog, ceramide synthase 5; LAG1 longevity assurance homolog 5; Sphingosine N-acyltransferase CERS5
Gene Aliases: CERS5; LASS5; Trh4
UniProt ID: (Human) Q8N5B7
Entrez Gene ID: (Human) 91012
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.