Invitrogen
Manufacturer Code:PA552250
Catalog # PIPA552250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Lass3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 3; CerS3; Dihydroceramide synthase 3; LAG1 homolog, ceramide synthase 3; LAG1 longevity assurance homolog 3; LASS3 CerS3 LAG1 homolog ceramide synthase 3 MGC27091 LAG1 longevity assurance homolog 3; Sphingosine N-acyltransferase CERS3
Gene Aliases: ARCI9; CERS3; LASS3
UniProt ID: (Human) Q8IU89
Entrez Gene ID: (Human) 204219
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.