Invitrogen
Manufacturer Code:MA527228
Catalog # PIMA527228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D9 |
Immunogen | Human recombit protein fragment corresponding to amino acids 62-136 of CERS2 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 2; Ceramide synthase 2 CerS2 FLJ10243 L3 MGC987 SP260 TMSG1 LASS2; CerS2; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; longevity assurance (LAG1, S. cerevisiae) homolog 2; SP260; Sphingosine N-acyltransferase CERS2; Tumor metastasis-suppressor gene 1 protein
Gene Aliases: CERS2; L3; LASS2; SP260; TMSG1
UniProt ID: (Human) Q96G23
Entrez Gene ID: (Human) 29956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.