Invitrogen
Manufacturer Code:PA578486
Catalog # PIPA578486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human LASS2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 2; CerS2; CERS2 FLJ10243 L3 LAG1 homolog ceramide synthase 2 LAG1 longevity assurance 2 LASS2 LASS2 MGC987 SP260 TMSG1 ceramide synthase 2 longevity assurance (LAG1 S. cerevisiae) homolog 2 tumor metastasis-suppressor gene 1 protein; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; longevity assurance (LAG1, S. cerevisiae) homolog 2; SP260; Sphingosine N-acyltransferase CERS2; Tumor metastasis-suppressor gene 1 protein
Gene Aliases: CERS2; L3; LASS2; SP260; TMSG1
UniProt ID: (Human) Q96G23
Entrez Gene ID: (Human) 29956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.