Invitrogen
Manufacturer Code:PA555560
Catalog # PIPA555560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Lass2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 2; Ceramide synthase 2 CerS2 FLJ10243 L3 MGC987 SP260 TMSG1 LASS2; CerS2; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; longevity assurance (LAG1, S. cerevisiae) homolog 2; longevity assurance homolog 2; SP260; Sphingosine N-acyltransferase CERS2; TRAM homolog 3; Translocating chain-associating membrane protein homolog 3; Tumor metastasis-suppressor gene 1 protein
Gene Aliases: 0610013I17Rik; AI225939; CERS2; L3; LASS2; SP260; TMSG1; TRH3
UniProt ID: (Human) Q96G23, (Mouse) Q924Z4
Entrez Gene ID: (Human) 29956, (Mouse) 76893, (Rat) 310667
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.