Invitrogen
Manufacturer Code:PA5112596
Catalog # PIPA5112596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Ceramide synthase 1 protein (1-52AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 1; Ceramide synthase 1 CERS1 EPM8 LAG1 LAG1 longevity assurance homolog 1 LASS1 longevity assurance (LAG1 S. cerevisiae) homolog 1 Longevity assurance gene 1 protein homolog 1 MGC90349 Protein UOG-1 UOG1 upstream of GDF1; CerS1; LAG1 homolog, ceramide synthase 1; LAG1 longevity assurance homolog 1; longevity assurance (LAG1, S. cerevisiae) homolog 1; Longevity assurance gene 1 protein homolog 1; longevity assurance homolog 1; longevity assurance-like protein 1; Protein UOG-1; upstream of GDF1
Gene Aliases: CERS1; EPM8; LAG1; LASS1; UOG1
UniProt ID: (Human) P27544
Entrez Gene ID: (Human) 10715, (Rat) 290658
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.