Invitrogen
Manufacturer Code:PA112918
Catalog # PIPA112918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the human Lass1 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ceramide synthase 1; CerS1; CerS1 LAG1 MGC90349 UOG1; LAG1 longevity assurance homolog 1; longevity assurance (LAG1, S. cerevisiae) homolog 1; Longevity assurance gene 1 protein homolog 1; Protein UOG-1; upstream of GDF1
Gene Aliases: CERS1; EPM8; LAG1; LASS1; UOG1
UniProt ID: (Human) P27544
Entrez Gene ID: (Human) 10715
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.