Invitrogen
Manufacturer Code:MA524646
Catalog # PIMA524646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL2980 |
Immunogen | Recombit protein corresponding to Human Laminin gamma-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alternatively spliced laminin gamma2 chain; B2T BM600 CSF EBR2 EBR2A LAMB2T LAMNB2 BM600-100kDa CSF 140 kDa subunit cell-scattering factor 140 kDa subunit epiligrin subunit gamma kalinin subunit gamma ladsin 140 kDa subunit laminin B2t chain laminin subunit gamma-2 large adhesive scatter factor 140 kDa subunit nicein subunit gamma; BM600-100kDa; Cell-scattering factor 140 kDa subunit; CSF 140 kDa subunit; Epiligrin subunit gamma; Kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; Laminin B2t chain; Laminin subunit gamma-2; laminin, gamma 2; Laminin-5 subunit gamma; Large adhesive scatter factor 140 kDa subunit; Nicein subunit gamma
Gene Aliases: B2T; BM600; CSF; EBR2; EBR2A; LAMB2T; LAMC2; LAMNB2
UniProt ID: (Human) Q13753
Entrez Gene ID: (Human) 3918
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.