Invitrogen
Manufacturer Code:PA579578
Catalog # PIPA579578
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human LAMC2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alternatively spliced laminin gamma2 chain; BM600-100kDa; Cell-scattering factor 140 kDa subunit; CSF 140 kDa subunit; Epiligrin subunit gamma; Kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; lamimin, gamma 2; Laminin B2t chain; laminin gamma2 chain; Laminin subunit gamma-2; Laminin subunit gamma-2 Cell-scattering factor 140 kDa subunit CSF 140 kDa subunit Epiligrin subunit gamma Kalinin subunit gamma Kalinin/nicein/epiligrin 100 kDa subunit Ladsin 140 kDa subunit Laminin B2t chain Laminin-5 subunit gamma Large adhesive scatter factor 140 kDa subunit Nicein subunit gamma LAMC2 LAMB2T LAMNB2; laminin, gamma 2; Laminin-5 subunit gamma; Large adhesive scatter factor 140 kDa subunit; Nicein subunit gamma; nicein, 100kD; nicein, 100kDa
Gene Aliases: AA589349; B2T; BM600; CSF; EBR2; EBR2A; LAMB2T; LAMC2; LAMNB2
UniProt ID: (Human) Q13753
Entrez Gene ID: (Human) 3918, (Mouse) 16782, (Rat) 192362
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.