Novus Biologicals
Manufacturer Code:NBP271115AF532
Catalog # NP27111A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3A2 |
Immunogen | Recombinant protein fragment corresponding to amino acids 424-773 of human Laminin beta 3 (NP_000219) produced in E.coli. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: BM600-125KDA Epiligrin subunit bata FLJ99565 Kalinin B1 chain Kalinin subunit beta kalinin-140kDa LAM5 Laminin 5 Laminin B1k chain laminin S B3 chain laminin subunit beta-3 laminin beta 3 laminin beta 3 (nicein (125kD) kalinin (140kD) BM600 (125kD)) Laminin-5 subunit beta LAMNB1 Nicein subunit beta nicein-125kDa; Epiligrin subunit bata; Kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; Laminin B1k chain; laminin beta 3; laminin S B3 chain; Laminin subunit beta-3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); Laminin-5 subunit beta; Nicein subunit beta; nicein-125kDa
Gene Aliases: AI1A; BM600-125KDA; LAM5; LAMB3; LAMNB1
UniProt ID: (Human) Q13751
Entrez Gene ID: (Human) 3914
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.