R&D Systems
Manufacturer Code:MAB4656SP
Catalog # MAB4656SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | AL-4 |
Immunogen | Purified fragment of chymotrypsin-digested mouse EHS tumor-derived Laminin-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: LAMA LAMA1 LAMA1 laminin alpha 1 S-LAM-alpha; Laminin A chain; Laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; S-LAM alpha; S-laminin subunit alpha
Gene Aliases: LAMA; LAMA1; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391
Entrez Gene ID: (Human) 284217
Molecular Function: extracellular matrix linker protein extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.