R&D Systems
Manufacturer Code:MAB4187100
Catalog # MAB4187100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 775016 |
Immunogen | <i>E. coli</i>-derived recombinant human Laminin alpha 1 Leu22-Met269 Accession &num P25391 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LAMA LAMA1 LAMA1 laminin alpha 1 S-LAM-alpha; Laminin A chain; Laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; S-LAM alpha; S-laminin subunit alpha
Gene Aliases: LAMA; LAMA1; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391
Entrez Gene ID: (Human) 284217
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.