Invitrogen
Manufacturer Code:MA121194
Catalog # PIMA121194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4C7 |
Immunogen | The A chain of human laminin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C do not freeze |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Laminin A chain; Laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; S-LAM alpha; S-laminin subunit alpha
Gene Aliases: LAMA; LAMA1; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391
Entrez Gene ID: (Human) 284217
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.