Invitrogen
Manufacturer Code:PA522901
Catalog # PIPA522901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Laminin isolated from mouse EHS tumor. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAMA S-LAM-alpha S-LAM alpha S-laminin subunit alpha laminin A chain laminin subunit alpha-1 laminin-3 subunit alpha; Laminin A chain; Laminin B1 chain; Laminin B2 chain; Laminin M chain; Laminin subunit alpha-1; Laminin subunit alpha-2; Laminin subunit beta-1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1 (formerly LAMB2); laminin-1; Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; Merosin heavy chain; S-LAM alpha; S-LAM gamma; S-laminin subunit alpha; S-laminin subunit gamma
Gene Aliases: AA408497; CLM; LAMA; Lama-1; LAMA1; LAMA2; LAMB1; LAMB2; LAMC1; LAMM; LIS5; PTBHS; S-LAM-alpha
UniProt ID: (Human) P25391, (Human) P24043, (Human) P07942, (Human) P11047, (Mouse) P19137
Entrez Gene ID: (Human) 284217, (Human) 3908, (Human) 3912, (Human) 3915, (Mouse) 16772, (Rat) 316758
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.