Novus Biologicals
Manufacturer Code:NBP272303G
Catalog # NBP272303G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C6 |
Immunogen | Full length recombinant protein of human Lamin B2 (NP_116126) produced in HEK293T cells. |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: lamin B2 lamin-B2 LMN2LAMB2 MGC2721; lamin B3; Lamin-B2
Gene Aliases: EPM9; LAMB2; LMN2; LMNB2
UniProt ID: (Human) Q14734
Entrez Gene ID: (Human) 84823
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.