Invitrogen
Manufacturer Code:MA332100A555
Catalog # PIMA33210AZ
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | E-3 |
Immunogen | Extracts from chicken nuclei |
Conjugate | Alexa Fluor 555 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: LAMB2 LMN2; lamin A/C; lamin B2; lamin B3; Lamin-B2; unnamed protein product
Gene Aliases: EPM9; LAMB2; lamin-B2; LMN2; LMNA; LMNB2; RGD1563803
UniProt ID: (Human) Q14734, (Chicken) P14732, (Mouse) P21619
Entrez Gene ID: (Human) 84823, (Chicken) 396222, (Mouse) 16907, (Rat) 299625
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.