Invitrogen
Manufacturer Code:PA519468
Catalog # PIPA519468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 400 - 500 of Mouse Lamin B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADLD LMN LMN2 LMNB MGC111419 lamin-B1; C-flanking peptide of NPY; CPON; Lamin-B1; Neuropeptide tyrosine; Neuropeptide Y; NPY; prepro-neuropeptide Y; Pro-neuropeptide Y
Gene Aliases: 0710005A05Rik; ADLD; lamin-b; Lamin-L(I); LMN; LMN2; LMNB; LMNB1; NPY; NPY02; PYY4; RATNPY; RATNPY02; XELAEV_18007951mg
UniProt ID: (Human) P20700, (Mouse) P14733, (Rat) P70615, (Human) P01303, (Mouse) P57774, (Rat) P07808
Entrez Gene ID: (Human) 4001, (Dog) 474663, (Mouse) 16906, (Rat) 116685, (Dog) 475257, (Human) 4852, (Mouse) 109648, (Rat) 24604, (Xenopus) 379745
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.