R&D Systems
Manufacturer Code:AF8525
Catalog # AF8525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant human Lamin B1 Met471-Ser582 Accession # P20700 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after opening. 6 months -20 to -70 °C under sterile conditions after |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADLD lamin B1 lamin-B1 LMN LMN2 LMNB LMNB1 MGC111419; Lamin-B1
Gene Aliases: ADLD; LMN; LMN2; LMNB; LMNB1
UniProt ID: (Human) P20700
Entrez Gene ID: (Human) 4001
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.