Novus Biologicals
Manufacturer Code:NBP271156
Catalog # NBP271156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F6 |
Immunogen | Full length human recombinant protein of human LMNA (NP_733821) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: 70 kDa lamin; CDCD1 CDDC CMD1A CMT2B1 dilated 1A (autosomal dominant) EMD2 FPLD HGPSFPL lamin A/C lamin A/C-like 1 lamin-A/C LDP1 LFP LGMD1B limb girdle muscular dystrophy 1B (autosomal dominant) LMN1IDC LMNC LMNL1 prelamin-A/C PRO170 kDa lamin progeria 1 (Hutchinson-Gilford type) renal carcinoma antigen NY-REN-32; lamin A/C-like 1; Lamin-A/C; Prelamin-A/C; Renal carcinoma antigen NY-REN-32
Gene Aliases: CDCD1; CDDC; CMD1A; CMT2B1; EMD2; FPL; FPLD; FPLD2; HGPS; IDC; LDP1; LFP; LGMD1B; LMN1; LMNA; LMNC; LMNL1; PRO1
UniProt ID: (Human) P02545
Entrez Gene ID: (Human) 4000
Molecular Function:
cytoskeletal protein
intermediate filament
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.