Invitrogen
Manufacturer Code:PA571304
Catalog # PIPA571304
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human IGLC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ig lambda chain V-I region BL2; immmunoglobulin lambda light chain variable region; immunoglobulin lambda light chain constant region; immunoglobulin lambda light chain V-J region; lambda-immunoglobulin
Gene Aliases: Clambda1; IgL; Igl-C1; IGLC; IGLC1; IGLL5
Entrez Gene ID: (Human) 3537, (Mouse) 110785, (Horse) 100060608
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.