Invitrogen
Manufacturer Code:MA140262
Catalog # 01-676-385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5F12.1.2 |
Immunogen | QWR region of Bovine Lactoferricin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Lactoferricin-B; Lactoferrin; Lactotransferrin; Lf; Lfcin-B
Gene Aliases: Lf; LTF
UniProt ID: (Bovine) P24627
Entrez Gene ID: (Bovine) 280846
Molecular Function: hydrolase protease serine protease transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.