Novus Biologicals
Manufacturer Code:NBP287714
Catalog # NB035336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human Lactase-like Protein/LCTL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ33279 KLG klotho gamma Klotho/lactase-phlorizin hydrolase-related protein KLPHKL lactase phlorizin hydrolase lactase-like lactase-like protein; KL lactase phlorizin hydrolase; klotho gamma; Klotho/lactase-phlorizin hydrolase-related protein; lactase-like; Lactase-like protein
Gene Aliases: KLG; KLPH; LCTL; UNQ3022/PRO9820
UniProt ID: (Human) Q6UWM7
Entrez Gene ID: (Human) 197021
Molecular Function:
glucosidase
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.