R&D Systems
Manufacturer Code:AF5984
Catalog # AF5984
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Lactase-like Protein/LCTL Arg23-Glu541 Accession # Q6UWM7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ33279 KLG klotho gamma Klotho/lactase-phlorizin hydrolase-related protein KLPH KLPHKL lactase phlorizin hydrolase Lactase like Protein lactase-like lactase-like protein LCTL; KL lactase phlorizin hydrolase; klotho gamma; Klotho/lactase-phlorizin hydrolase-related protein; lactase-like; Lactase-like protein
Gene Aliases: KLG; KLPH; LCTL; UNQ3022/PRO9820
UniProt ID: (Human) Q6UWM7
Entrez Gene ID: (Human) 197021
Molecular Function:
glucosidase
glycosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.