Novus Biologicals
Manufacturer Code:NB10062705
Catalog # NB10062705
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Lactalbumin from human milk |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-lactalbumin; alpha-lactalbumin lactalbumin alpha- Lactose synthase B protein Lysozyme-like protein 7 LYZL7 MGC138521 MGC138523; lactalbumin, alpha-; Lactose synthase B protein; lysozyme G; Lysozyme-like protein 7
Gene Aliases: LALBA; LYZG; LYZL7
UniProt ID: (Human) P00709
Entrez Gene ID: (Human) 3906
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.