Invitrogen
Manufacturer Code:PA535130
Catalog # PIPA535130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 77-105 amino acids from the N-terminal region of human LZTS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: F37 esophageal cancer-related leucine zipper motif 1; F37/Esophageal cancer-related gene-coding leucine-zipper motif; Fez1; Leucine zipper putative tumor suppressor 1; leucine zipper, putative tumor suppressor 1; leucine-zipper protein FEZ1; LZTS1 FEZ1 Leucine zipper putative tumor suppressor 1 F37/esophageal cancer-related gene-coding leucine-zipper motif Fez1
Gene Aliases: F37; FEZ1; LZTS1
UniProt ID: (Human) Q9Y250, (Mouse) P60853
Entrez Gene ID: (Human) 11178, (Mouse) 211134
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.