Invitrogen
Manufacturer Code:PA520903
Catalog # PIPA520903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the carboxy terminus of human LZTR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Leucine zipper transcription regulator 2; LZTR2 PGPR-p117 RGPR SEC16S RGPR-p117 leucine zipper transcription regulator 2 protein SEC16 homolog B protein transport protein Sec16B regucalcin gene promoter region-related protein p117 regucalcin gene promotor region related protein; protein SEC16 homolog B; Protein transport protein Sec16B; Regucalcin gene promoter region-related protein p117; regucalcin gene promotor region related protein; RGPR-p117; SEC16 homolog B
Gene Aliases: KIAA1928; LZTR2; PGPR-p117; RGPR; Rgpr-p117; SEC16B; SEC16S
UniProt ID: (Human) Q96JE7, (Mouse) Q91XT4, (Rat) Q75N33
Entrez Gene ID: (Human) 89866, (Mouse) 89867, (Rat) 89868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.