Novus Biologicals
Manufacturer Code:NBP217194
Catalog # NBP217194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human LZTR2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686C2486 FLJ23871 FLJ25761 KIAA1928 Leucine zipper transcription regulator 2FLJ36620 LZTR2SEC16Sprotein SEC16 homolog B PGPR-p117 protein transport protein Sec16B Regucalcin gene promoter region-related protein p117 regucalcin gene promotor region related protein RGPRFLJ33652 RGPR-p117 SEC16 homolog B SEC16 homolog B (S. cerevisiae); Leucine zipper transcription regulator 2; protein SEC16 homolog B; Protein transport protein Sec16B; Regucalcin gene promoter region-related protein p117; regucalcin gene promotor region related protein; RGPR-p117; SEC16 homolog B
Gene Aliases: KIAA1928; LZTR2; PGPR-p117; RGPR; SEC16B; SEC16S
UniProt ID: (Human) Q96JE7
Entrez Gene ID: (Human) 89866
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.